Tetramethyl methylenebisphosphonate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetramethyl methylenebis(phosphonate), Tetramethyl methylenediphosphonate
A13441-06
A13441-14
A13441-22
CAAAA13441-06EA
211.72
CAD
CAAAA13441-06
CAAAA13441-14
CAAAA13441-22
Tetramethyl methylenebisphosphonate 98+%
Tetramethyl methylenebisphosphonate
Miscible in water
Purity: 98+%
CAS No.: 16001-93-7
Molecular Formula: C5H14O6P2
Formula Weight: 232.11
Refractive Index: 1.4550
Boiling Point: 152-154°/2.5mm
MDL No.: MFCD00014884
Formula:
C₅H₁₄O₆P₂ Boiling Pt: 152…154 °C/2.5 mm Storage Temperature: Ambient |
MDL Number:
MFCD00014884 CAS Number: 16001-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...