Methyl-4-phenylbenzoate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: p-Phenylbenzoic acid-OMe, Methyl biphenyl-4-carboxylate, Methyl [1,1'-biphenyl]-4-carboxylate

A13430-06 A13430-18
CAAAAA13430-06EA 62.57 CAD
CAAAAA13430-06 CAAAA13430-18 CAAAA13430-30
Methyl-4-phenylbenzoate 98+%
Methyl 4-phenylbenzoate

Purity: 98+% CAS No.: 720-75-2 Molecular Formula: C14H12O2 Formula Weight: 212.25 Odor: Odorless Melting Point: 116-119° Beilstein Reference: 1954046 MDL No.: MFCD00017200

Formula: C₁₄H₁₂O₂
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00017200
CAS Number: 720-75-2
EINECS: 211-954-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR