5-Azacytosine, (max. 7% H₂O) 98% (dry weight)
Supplier: Thermo Scientific Chemicals
A13410-06
A13410-14
CAAAA13410-06EA
66.52
CAD
CAAAA13410-06
CAAAA13410-14
5-Azacytosine, (max. 7% H₂O) 98% (dry weight)
5-Azacytosine
Purity: 98%
CAS No.: 931-86-2
Molecular Formula: C3H4N4O
Formula Weight: 112.09
Melting Point:
Formula:
C₃H₄N₄O Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149402 CAS Number: 931-86-2 EINECS: 213-242-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...