4-Hydrazinobenzenesulfonic acid hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

A13362-06 A13362-14 A13362-22
CAAAA13362-06EA 56.56 CAD
CAAAA13362-06 CAAAA13362-14 CAAAA13362-22
4-Hydrazinobenzenesulfonic acid hydrate 98%
4-Hydrazinobenzenesulfonic acid hydrate

Purity: 98% CAS No.: 98-71-5 Molecular Formula: C6H8N2O3S.xH2O Formula Weight: 188.20(anhy) Melting Point: ca 286° dec. Beilstein Reference: 514779 MDL No.: MFCD00025097

Formula: C₆H₈N₂O₃S·xH₂O
MW: 188.20 g/mol (anhy)
Melting Pt: ∼ 286 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00025097
CAS Number: 98-71-5
EINECS: 202-694-2
Merck Index: 14,4773

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR