4-Bromophenyl octyl ether 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Bromo-4-(n-octyloxy)benzene

A13334-03 A13334-14
CAAAA13334-03EA 56.41 CAD
CAAAA13334-03 CAAAA13334-06 CAAAA13334-14
4-Bromophenyl octyl ether 98%
4-Bromophenyl octyl ether

Purity: 98% CAS No.: 96693-05-9 Molecular Formula: C14H21BrO Formula Weight: 285.23 Refractive Index: 1.5180 Boiling Point: 135-137°/0.5mm MDL No.: MFCD00043445

Formula: C₁₄H₂₁BrO
MW: 285.23 g/mol
Boiling Pt: 135…137 °C/0.5 mm
Storage Temperature: Ambient
MDL Number: MFCD00043445
CAS Number: 96693-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR