4-Chloro-DL-phenylalanine 98+%
Supplier: Thermo Scientific Chemicals
A13323-09
A13323-18
CAAAA13323-09EA
152.63
CAD
CAAAA13323-09
CAAAA13323-18
4-Chloro-DL-phenylalanine 98+%
4-Chloro-DL-phenylalanine
Purity: 98+%
CAS No.: 7424-00-2
Molecular Formula: C9H10ClNO2
Formula Weight: 199.64
Melting Point: ca 265° dec.
Beilstein Reference: 2805758
Formula:
C₉H₁₀ClNO₂ Melting Pt: ∼ 265 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002601 CAS Number: 7424-00-2 EINECS: 231-051-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...