2-Sulfobenzoic cyclic anhydride 94%

Supplier: Thermo Scientific Chemicals
Danger

A13314-14 A13314-22 A13314-36
CAAAA13314-14EA 115.5 CAD
CAAAA13314-14 CAAAA13314-22 CAAAA13314-36
2-Sulfobenzoic cyclic anhydride 94%
2-Sulphobenzoic cyclic anhydride

Purity: 94% CAS No.: 81-08-3 Molecular Formula: C7H4O4S Formula Weight: 184.17 Melting Point: ca 120° Boiling Point: 184-186°/18mm MDL No.: MFCD00005879

Formula: C₇H₄O₄S
Boiling Pt: 184…186 °C/18 mm
Melting Pt: ∼ 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00005879
CAS Number: 81-08-3
EINECS: 201-322-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR