3-Methyl-2-thiophenecarbonitrile 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13305-14
A13305-03
A13305-06
CAAAAA13305-14EA
560.9
CAD
CAAAAA13305-14
CAAAA13305-03
CAAAA13305-06
3-Methyl-2-thiophenecarbonitrile 96%
3-Methyl-2-thiophenecarbonitrile
Purity: 96%
CAS No.: 55406-13-8
Molecular Formula: C6H5NS
Formula Weight: 123.18
Refractive Index: 1.5530
Boiling Point: 91-93°/12mm
UN No.: 3276
MDL No.: MFCD00052086
Formula:
C₆H₅NS Boiling Pt: 91…93 °C/12 mm Storage Temperature: Ambient |
MDL Number:
MFCD00052086 CAS Number: 55406-13-8 UN: 3276 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...