Trimethyl phosphonoacetate 98%

Supplier: Thermo Scientific Chemicals
Warning

A13301-14 A13301-22 A13301-36
CAAAA13301-14EA 76.16 CAD
CAAAA13301-14 CAAAA13301-22 CAAAA13301-36
Trimethyl phosphonoacetate 98%
Trimethyl phosphonoacetate

Fieser: 2,442 7,394


Purity: 98% CAS No.: 5927-18-4 Molecular Formula: C5H11O5P Formula Weight: 182.11 Flash Point: 70°(158°F) Refractive Index: 1.4370 Boiling Point: 119-120°/1mm Beilstein Reference: 1865177 MDL No.: MFCD00008452

Formula: C₅H₁₁O₅P
Boiling Pt: 119…120 °C (1 mmHg)
Density: 1.263
Flash Pt: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008452
CAS Number: 5927-18-4
EINECS: 227-663-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR