Trifluoroacetaldehyde hydrate ≥75% in water, tech.

Supplier: Thermo Scientific Chemicals
Danger

A13234-09 A13234-18
CAAAA13234-09EA 159.58 CAD
CAAAA13234-09 CAAAA13234-18
Trifluoroacetaldehyde hydrate ≥75% in water, tech.
Trifluoroacetaldehyde hydrate

Purity: 75% CAS No.: 421-53-4 Molecular Formula: C2H3F3O2 Formula Weight: 116.04(anhy) Flash Point: None Boiling Point: 104-106° UN No.: 1989 MDL No.: MFCD00167359

Formula: C₂H₃F₃O₂
Boiling Pt: 104…106 °C
Density: 1.44
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00167359
CAS Number: 421-53-4
EINECS: 207-006-4
UN: 1989
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR