2-Nonenoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13228-22
A13228-14
CAAAAA13228-22EA
559.32
CAD
CAAAAA13228-22
CAAAA13228-14
2-Nonenoic acid 98%
2-Nonenoic acid
Purity: 98%
CAS No.: 3760-11-0
Molecular Formula: C9H16O2
Formula Weight: 156.23
Flash Point:
Formula:
C₉H₁₆O₂ Boiling Pt: 130…132 °C/2 mm Density: 0.93 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014020 CAS Number: 3760-11-0 EINECS: 223-171-5 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...