4-(1H-Imidazol-1-yl)phenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13211-03
A13211-14
CAAAAA13211-03EA
47.4
CAD
CAAAAA13211-03
CAAAA13211-14
4-(1H-Imidazol-1-yl)phenol 97%
4-(1H-Imidazol-1-yl)phenol
Purity: 97%
CAS No.: 10041-02-8
Molecular Formula: C9H8N2O
Formula Weight: 160.18
Melting Point: 204-208°
Beilstein Reference: 509999
Formula:
C₉H₈N₂O Melting Pt: 204…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005281 CAS Number: 10041-02-8 EINECS: 233-121-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...