N-Hydroxy-5-norbornene-2,3-dicarboximide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: endo-N-Hydroxybicyclo[2.2.1]hept-5-ene-2,3-dicarboximide

A13205-09 A13205-18 A13205-30
CAAAA13205-09EA 65.89 CAD
CAAAA13205-09 CAAAA13205-18 CAAAA13205-30
N-Hydroxy-5-norbornene-2,3-dicarboximide 97%
N-Hydroxy-5-norbornene-2,3-dicarboximide

Purity: 97% CAS No.: 21715-90-2 Molecular Formula: C9H9NO3 Formula Weight: 179.18 Melting Point: 166-170° Beilstein Reference: 13540 MDL No.: MFCD00065691

Formula: C₉H₉NO₃
MW: 179.18 g/mol
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00167583
CAS Number: 21715-90-2
EINECS: 244-538-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR