Magnesium hexafluorosilicate hexahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A13189-0C A13189-0E A13189-36
CAAAAA13189-0CEA 1044.38 CAD
CAAAAA13189-0C CAAAAA13189-0E CAAAAA13189-36
Magnesium hexafluorosilicate hexahydrate 98%
Magnesium hexafluorosilicate hexahydrate

Purity: 98% CAS No.: 18972-56-0 Molecular Formula: MgSiF6.6H2O Formula Weight: 274.47 (166.39anhy) Odor: Odorless UN No.: 2853 MDL No.: MFCD00150250

Formula: F₆MgSi·6H₂O
Density: 1.79
Storage Temperature: Ambient
MDL Number: MFCD00150250
CAS Number: 18972-56-0
EINECS: 241-022-2
UN: 2853
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR