2-Mercapto-4-methylpyrimidine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Methylpyrimidine-2(1H)-thione hydrochloride
A13145-14
A13145-22
CAAAA13145-14EA
58.93
CAD
CAAAA13145-14
CAAAA13145-22
2-Mercapto-4-methylpyrimidine hydrochloride 99%
2-Mercapto-4-methylpyrimidine hydrochloride
Purity: 99%
CAS No.: 6959-66-6
Molecular Formula: C5H6N2S.HCl
Formula Weight: 162.64
Melting Point: ca 265° dec.
Beilstein Reference: 3691520
MDL No.: MFCD00012783
Formula:
C₅H₇ClN₂S MW: 162.635 g/mol Melting Pt: ∼ 265 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012783 CAS Number: 6959-66-6 EINECS: 230-152-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...