Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide

A13143-03 A13143-06 A13143-14
CAAAA13143-03EA 50.56 CAD
CAAAA13143-03 CAAAA13143-06 CAAAA13143-14
Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide) 98%
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimide)

Purity: 98% CAS No.: 82911-69-1 Molecular Formula: C19H15NO5 Formula Weight: 337.34 Storage Temperature: 2°C to 8°C Melting Point: 150-153° Beilstein Reference: 3569540 UN No.: 3077 MDL No.: MFCD00010733

Formula: C₁₉H₁₅NO₅
MW: 337.3 g/mol
Melting Pt: 150…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010733
CAS Number: 82911-69-1
EINECS: 433-520-5
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR