L(-)-Arabitol 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
L-(-)-Arabinitol, (2S,4S)-Pentane-1,2,3,4,5-pentaol, L-(-)-Arabitol, L-Arabinitol
A13103-22
A13103-06
A13103-14
CAAAAA13103-22EA
856.36
CAD
CAAAAA13103-22
CAAAA13103-06
CAAAA13103-14
L(-)-Arabitol 98%
L(-)-Arabitol
Purity: 98%
CAS No.: 7643-75-6
Molecular Formula: C5H12O5
Formula Weight: 152.15
Melting Point: 98-103°
Beilstein Reference: 1720521
Merck Reference: 14,762
Optical Rotation: -11° (c=5 in 8% borax soln)
MDL No.: MFCD00064290
Formula:
C₅H₁₂O₅ Melting Pt: 98…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064290 CAS Number: 7643-75-6 EINECS: 231-582-6 Merck Index: 14,762 |
Specification Test Results
Optical rotation | -11° (c=5 in 8% borax soln) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...