Methyl-1-methyl-1H-pyrrole-2-carboxylate 99%

Supplier: Thermo Scientific Chemicals

A13095-06 A13095-14
CAAAA13095-06EA 183.28 CAD
CAAAA13095-06 CAAAA13095-14
Methyl-1-methyl-1H-pyrrole-2-carboxylate 99%
Methyl-1-methyl-1H-pyrrole-2-carboxylate

Purity: 99% CAS No.: 37619-24-2 Molecular Formula: C7H9NO2 Formula Weight: 139.16 Refractive Index: 1.5220 Boiling Point: 96-98°/15mm Beilstein Reference: 116477 MDL No.: MFCD00052747

Formula: C₇H₉NO₂
Boiling Pt: 96…98 °C/15 mm
Storage Temperature: Ambient
MDL Number: MFCD00052747
CAS Number: 37619-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR