2-(1-Piperidinyl)aniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13073-06
A13073-14
CAAAAA13073-22EA
2539.63
CAD
CAAAAA13073-22
CAAAA13073-06
CAAAA13073-14
2-(1-Piperidinyl)aniline 98%
2-(1-Piperidinyl)aniline
Purity: 98%
CAS No.: 39643-31-7
Molecular Formula: C11H16N2
Formula Weight: 176.26
Melting Point: 44-47°
Boiling Point: 96-98°/1mm
UN No.: 2811
MDL No.: MFCD00047467
Formula:
C₁₁H₁₆N₂ Boiling Pt: 96…98 °C/1 mm Melting Pt: 44…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047467 CAS Number: 39643-31-7 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...