Sodium-2-naphthalenesulfonate 98% may cont. up to 10% residual inorganic salts and water

Supplier: Thermo Scientific Chemicals
Warning

A13066-0B A13066-30
CAAAA13066-0BEA 214.88 CAD
CAAAA13066-0B CAAAA13066-30
Sodium-2-naphthalenesulfonate 98% may cont. up to 10% residual inorganic salts and water
Sodium-2-naphthalenesulfonate

Purity: 98% CAS No.: 532-02-5 Molecular Formula: C10H7NaO3S Formula Weight: 230.22 Beilstein Reference: 3659024 MDL No.: MFCD00064186

Formula: C₁₀H₈O₃S
Storage Temperature: Ambient
MDL Number: MFCD00064186
CAS Number: 532-02-5
EINECS: 208-523-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR