3-(2-Furyl)acrolein 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13042-06
A13042-14
CAAAA13042-06EA
51.03
CAD
CAAAA13042-06
CAAAA13042-14
3-(2-Furyl)acrolein 99%
3-(2-Furyl)acrolein
Purity: 99%
CAS No.: 623-30-3
Molecular Formula: C7H6O2
Formula Weight: 122.12
Storage Temperature: 2°C to 8°C
Flash Point: 99°(210°F)
Melting Point: 49-52°
Boiling Point: 95°/5mm
Beilstein Reference: 107570
UN No.: 1759
MDL No.: MFCD00003256
Formula:
C₇H₆O₂ Boiling Pt: 95 °C/5 mm Melting Pt: 49…52 °C Flash Pt: 99 °C(210 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003256 CAS Number: 623-30-3 EINECS: 210-785-3 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...