1,3-dimethyl-1H-pyrazol-5-amine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Amino-1,3-dimethylpyrazole, 5-Amino-1,3-dimethyl-1H-pyrazole

A12980-03 A12980-06 A12980-14
CAAAA12980-03EA 47.4 CAD
CAAAA12980-03 CAAAA12980-06 CAAAA12980-14
1,3-dimethyl-1H-pyrazol-5-amine 98%
1,3-dimethyl-1H-pyrazol-5-amine

Purity: 98% CAS No.: 3524-32-1 Molecular Formula: C5H9N3 Formula Weight: 111.14 Flash Point: 88°(190°F) Melting Point: 72-77° Beilstein Reference: 113420 MDL No.: MFCD00051651

Formula: C₅H₉N₃
Melting Pt: 72…77 °C
Flash Pt: 88 °C(190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051651
CAS Number: 3524-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR