4-Acetoxycinnamic acid predominantly trans ≥98%

Supplier: Thermo Scientific Chemicals
Warning

A12979-06 A12979-22
CAAAA12979-06EA 67.31 CAD
CAAAA12979-06 CAAAA12979-14 CAAAA12979-22
4-Acetoxycinnamic acid predominantly trans ≥98%
4-Acetoxycinnamic acid

Purity: 98+% CAS No.: 15486-19-8 Molecular Formula: C11H10O4 Formula Weight: 206.20 Odor: Odorless Melting Point: 205-208° Beilstein Reference: 2645987 MDL No.: MFCD00016847

Formula: C₁₁H₁₀O₄
Melting Pt: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00016847
CAS Number: 15486-19-8
EINECS: 239-512-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR