Reactive blue 4 (dye content) 35%, dark blue powder C.I. 61205
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12911-14
A12911-22
CAAAA12911-14EA
89.27
CAD
CAAAA12911-14
CAAAA12911-22
Reactive blue 4 (dye content) 35%, dark blue powder C.I. 61205
Reactive blue 4
CAS No.: 13324-20-4
Molecular Formula: C23H14Cl2N6O8S2
Formula Weight: 637.44
Odor: Odorless
MDL No.: MFCD00001218
Formula:
C₂₃H₁₄Cl₂N₆O₈S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00001218 CAS Number: 13324-20-4 EINECS: 236-363-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...