Ethyl-2-(triphenylphosphoranylidene)acetate ≥98%

Supplier: Thermo Scientific Chemicals

A12896-09 A12896-18 A12896-30
CAAAA12896-09EA 57.51 CAD
CAAAA12896-09 CAAAA12896-18 CAAAA12896-30
Ethyl-2-(triphenylphosphoranylidene)acetate ≥98%
Ethyl-2-(triphenylphosphoranylidene)acetate

Fieser: 1,112 2,60 10,78 15,157


Purity: 98+% CAS No.: 1099-45-2 Molecular Formula: CH3CH2O2CCH=P(C6H5)3 Formula Weight: 348.38 Storage Temperature: 2°C to 8°C Melting Point: 126-130° Beilstein Reference: 754639 MDL No.: MFCD00009183

Formula: C₂₂H₂₁O₂P
Melting Pt: 126…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009183
CAS Number: 1099-45-2
EINECS: 214-151-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR