3-Isocyanatopropyltriethoxysilane 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
ICPTES, Triethoxy(3-isocyanatopropyl)silane, (3-Isocyanatopropyl)triethoxysilane, Isocyanic acid 3-(Triethoxysilyl)propyl ester, 3-(Triethoxysilyl)propyl Isocyanate
A12890-06
A12890-14
CAAAA12890-06EA
72.68
CAD
CAAAA12890-06
CAAAA12890-14
3-Isocyanatopropyltriethoxysilane 95%
3-Isocyanatopropyltriethoxysilane
Purity: 95%
CAS No.: 24801-88-5
Molecular Formula: C10H21NO4Si
Formula Weight: 247.37
Flash Point: 77°(170°F)
Refractive Index: 1.4210
Odor: Mild
Boiling Point: 238°
Beilstein Reference: 4404652
UN No.: 2927
MDL No.: MFCD00051459
Formula:
C₁₀H₂₁NO₄Si MW: 247.366 g/mol Boiling Pt: 238 °C Density: 0.990 Flash Pt: 77 °C (170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051459 CAS Number: 24801-88-5 EINECS: 246-467-6 UN: 2927 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...