Tris[4-(trifluoromethyl)phenyl]phosphine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(4-trifluoromethylphenyl)phosphine, Tris(4-(trifluoromethyl)phenyl)phosphine

A12885-03 A12885-06
CAAAA12885-03EA 224.36 CAD
CAAAA12885-03 CAAAA12885-06
Tris[4-(trifluoromethyl)phenyl]phosphine 98%
Tris[4-(trifluoromethyl)phenyl]phosphine

Purity: 98% CAS No.: 13406-29-6 Molecular Formula: C21H12F9P Formula Weight: 466.29 Melting Point: 72-75° MDL No.: MFCD00058883

Formula: C₂₁H₁₂F₉P
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00058883
CAS Number: 13406-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR