Dinonyl phthalate (mixture of isomers) 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Phthalic acid dinonyl ester

A12871-09 A12871-18 A12871-30
CAAAA12871-09EA 73.94 CAD
CAAAA12871-09 CAAAA12871-18 CAAAA12871-30
Dinonyl phthalate (mixture of isomers) 98%
Dinonyl phthalate (mixture of isomers)

Purity: 98% CAS No.: 84-76-4 Molecular Formula: C26H42O4 Formula Weight: 418.62 Flash Point: 216°(421°F)

Formula: C₂₆H₄₂O₄
MW: 418.62 g/mol
Density: 0.97
Flash Pt: 216 °C(421 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036237
CAS Number: 84-76-4
EINECS: 201-560-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR