p-Toluenesulfonylacetonitrile 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [(4-Methylphenyl)sulfonyl]acetonitrile, [(4-Methylphenyl)sulphonyl]acetonitrile

A12857-06 A12857-14 A12857-22
CAAAA12857-06EA 43.61 CAD
CAAAA12857-06 CAAAA12857-14 CAAAA12857-22
p-Toluenesulfonylacetonitrile 98+%
p-Toluenesulphonylacetonitrile

Purity: 98+% CAS No.: 5697-44-9 Molecular Formula: C9H9NO2S Formula Weight: 195.24 Melting Point: 147-152° Beilstein Reference: 1842131 UN No.: 3439 MDL No.: MFCD00039487

Formula: C₉H₉NO₂S
Melting Pt: 147…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00039487
CAS Number: 5697-44-9
EINECS: 227-173-7
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR