1-Benzylpiperidin-3-one hydrochloride hydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12849-06
A12849-14
CAAAAA12849-03EA
107.79
CAD
CAAAAA12849-03
CAAAAA12849-06
CAAAAA12849-14
1-Benzylpiperidin-3-one hydrochloride hydrate
1-Benzylpiperidin-3-one hydrochloride hydrate
CAS No.: 50606-58-1
Molecular Formula: C12H15NO.HCl.xH2O
Formula Weight: 225.72(anhy)
Storage Temperature: 2°C to 8°C
Melting Point: 105-107°
Beilstein Reference: 3705146
MDL No.: MFCD00150096
Formula:
C₁₂H₁₅NO.HCl.xH₂O Melting Pt: 105…107 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00150096 CAS Number: 346694-73-3 EINECS: 256-646-0 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...