4-Octyloxybenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12843-22
A12843-06
A12843-14
CAAAAA12843-22EA
521.4
CAD
CAAAAA12843-22
CAAAA12843-03
CAAAA12843-06
CAAAA12843-14
4-Octyloxybenzonitrile 98%
4-Octyloxybenzonitrile
Purity: 98%
CAS No.: 88374-55-4
Molecular Formula: C15H21NO
Formula Weight: 231.34
Refractive Index: 1.5100
Melting Point: 24-26°
Boiling Point: 171-173°/2mm
Beilstein Reference: 2844884
UN No.: 3439
MDL No.: MFCD00043483
Formula:
C₁₅H₂₁NO Boiling Pt: 171…173 °C/2 mm Melting Pt: 24…26 °C Density: 0.937 Storage Temperature: Ambient |
MDL Number:
MFCD00043483 CAS Number: 88374-55-4 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...