Ammonium copper(II) sulfate hexahydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

A12834-0B A12834-30
CAAAAA12834-0BEA 319.16 CAD
CAAAAA12834-0B CAAAAA12834-30
Ammonium copper(II) sulfate hexahydrate 99%
Ammonium copper(II) sulfate hexahydrate

Purity: 99% CAS No.: 13587-26-3 Molecular Formula: (NH4)2Cu(SO4)2.6H2O Formula Weight: 399.83 (291.75anhy) MDL No.: MFCD00150440

Formula: (NH₄)₂Cu(SO₄)₂·6H₂O
MW: 399.83 g/mol (291.75 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150440
CAS Number: 13587-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR