Thiophen-3-acetonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12805-06
A12805-14
CAAAA12805-06EA
131.93
CAD
CAAAA12805-06
CAAAA12805-14
Thiophen-3-acetonitrile 98%
Thiophen-3-acetonitrile
Purity: 98%
CAS No.: 13781-53-8
Molecular Formula: C6H5NS
Formula Weight: 123.18
Flash Point: 119°(246°F)
Refractive Index: 1.5430
Boiling Point: 240-242°
Beilstein Reference: 107774
UN No.: 3276
MDL No.: MFCD00005471
Formula:
C₆H₅NS Boiling Pt: 240…242 °C Density: 1.080 Flash Pt: 119 °C (246 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005471 CAS Number: 13781-53-8 EINECS: 237-433-1 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...