Di-tert-butyl malonate 98+% stabilised

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with potassium carbonate

A12774-09 A12774-18 A12774-30
CAAAA12774-09EA 105.7 CAD
CAAAA12774-09 CAAAA12774-18 CAAAA12774-30
Di-tert-butyl malonate 98+% stabilised
Di-tert-butyl malonate

Fieser: 1,210


Purity: 98+% CAS No.: 541-16-2 Molecular Formula: C11H20O4 Formula Weight: 216.28 Flash Point: 88°(190°F) Refractive Index: 1.4190 Melting Point: -6° Boiling Point: 65-67°/1mm Beilstein Reference: 1781766 Merck Reference: 14,3034 MDL No.: MFCD00008810

Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 65…67 °C (1 mmHg)
Melting Pt: –6 °C
Density: 0.965
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008810
CAS Number: 541-16-2
EINECS: 208-769-6
Merck Index: 14,3034

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR