Diethyl mesoxalate (diethyl ketomalonate) 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diethyl 2-oxomalonate
A12771-06
A12771-14
A12771-22
CAAAA12771-06EA
73.47
CAD
CAAAA12771-06
CAAAA12771-14
CAAAA12771-22
Diethyl mesoxalate (diethyl ketomalonate) 95%
Diethyl mesoxalate (Diethyl ketomalonate)
Fieser: 6,188 8,170 10,143
Purity: 95%
CAS No.: 609-09-6
Molecular Formula: C7H10O5
Formula Weight: 174.15
Flash Point:
Formula:
C₇H₁₀O₅ Boiling Pt: 208…210 °C Melting Pt: ca …70 °C Density: 1.142 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009121 CAS Number: 609-09-6 EINECS: 210-176-2 Merck Index: 14,5913 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...