DL-Phenylalanine methyl ester hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
|
Synonyms:
H-DL-Phe-OMe·HCl, Methyl 2-amino-3-phenylpropanoate hydrochloride
A12758-06
A12758-14
A12758-22
CAAAA12758-06EA
90.53
CAD
CAAAA12758-06
CAAAA12758-14
CAAAA12758-22
DL-Phenylalanine methyl ester hydrochloride 98%
DL-Phenylalanine methyl ester hydrochloride
Purity: 98%
CAS No.: 5619-07-8
Molecular Formula: C10H13NO2.HCl
Formula Weight: 215.68
Melting Point: ca 160° dec.
Beilstein Reference: 3597947
MDL No.: MFCD00066113
Formula:
C₁₀H₁₄ClNO₂ MW: 215.677 g/mol Melting Pt: ∼ 160 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00066113 CAS Number: 5619-07-8 EINECS: 227-049-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...