4-Methoxy-β-nitrostyrene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methoxy-4-(2-nitrovinyl)benzene, trans-4-Methoxy-β-nitrostyrene

A12745-22 A12745-06 A12745-14
CAAAAA12745-22EA 1635.3 CAD
CAAAAA12745-22 CAAAA12745-06 CAAAA12745-14
4-Methoxy-β-nitrostyrene 98%
4-Methoxy-β-nitrostyrene

Purity: 98% CAS No.: 5576-97-6 Molecular Formula: C9H9NO3 Formula Weight: 179.18 Storage Temperature: 2°C to 8°C Melting Point: 86-88° Beilstein Reference: 2047935 MDL No.: MFCD00024826

Formula: C₉H₉NO₃
MW: 179.173 g/mol
Melting Pt: 86…88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00024826
CAS Number: 3179-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR