Ethyl succinyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12734-14
A12734-22
A12734-36
CAAAA12734-14EA
109.97
CAD
CAAAA12734-14
CAAAA12734-22
CAAAA12734-36
Ethyl succinyl chloride 97%
Ethyl succinyl chloride
Purity: 97%
CAS No.: 14794-31-1
Molecular Formula: ClOCCH2CH2CO2CH2CH3
Formula Weight: 164.59
Flash Point: 84°(183°F)
Refractive Index: 1.4370
Odor: Acrid
Boiling Point: 88-90°/11mm
Beilstein Reference: 1766126
UN No.: 3265
MDL No.: MFCD00000751
Formula:
C₆H₉ClO₃ Boiling Pt: 88…90 °C/11 mm Density: 1.163 Flash Pt: 84 °C (183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000751 CAS Number: 14794-31-1 EINECS: 238-855-9 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...