Procion blue H-5R
Supplier: Thermo Scientific Chemicals
A12719-14
A12719-22
CAAAA12719-14EA
71.42
CAD
CAAAA12719-14
CAAAA12719-22
Procion blue H-5R
Procion blue H-5R
CAS No.: 12236-84-9
MDL No.: MFCD00070851
Storage Temperature:
Ambient |
MDL Number:
MFCD00070851 CAS Number: 12236-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...