3-Amino-5-phenyl-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-5-phenylpyrazole

A12701-06 A12701-14
CAAAA12701-03EA 103.93 CAD
CAAAA12701-03 CAAAA12701-06 CAAAA12701-14
3-Amino-5-phenyl-1H-pyrazole 98%
3-Amino-5-phenyl-1H-pyrazole

Purity: 98% CAS No.: 1572-10-7 Molecular Formula: C9H9N3 Formula Weight: 159.19 Melting Point: 124-128° Beilstein Reference: 4947

Formula: C₉H₉N₃
MW: 159.19 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00191749
CAS Number: 1572-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR