Methyl-2-thiophenecarboxylate 97%
Supplier: Thermo Scientific Chemicals
A12677-22
A12677-06
A12677-14
CAAAAA12677-22EA
952.74
CAD
CAAAAA12677-22
CAAAA12677-06
CAAAA12677-14
Methyl-2-thiophenecarboxylate 97%
Methyl-2-thiophenecarboxylate
Purity: 97%
CAS No.: 5380-42-7
Molecular Formula: C6H6O2S
Formula Weight: 142.18
Refractive Index: 1.5420
Boiling Point: 95-97°/15mm
Beilstein Reference: 111111
MDL No.: MFCD00016895
Formula:
C₆H₆O₂S Boiling Pt: 95…97 °C/15 mm Density: 1.230 Storage Temperature: Ambient |
MDL Number:
MFCD00016895 CAS Number: 5380-42-7 EINECS: 226-371-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...