(3-Phenylpropyl)triphenylphosphonium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12669-09
A12669-18
CAAAA12669-09EA
81.21
CAD
CAAAA12669-09
CAAAA12669-18
CAAAA12669-30
(3-Phenylpropyl)triphenylphosphonium bromide 98%
(3-Phenylpropyl)triphenylphosphonium bromide
Purity: 98%
CAS No.: 7484-37-9
Molecular Formula: C27H26BrP
Formula Weight: 461.39
Odor: Odorless
Melting Point: 208-212°
Beilstein Reference: 4173336
MDL No.: MFCD00051886
Formula:
C₂₇H₂₆BrP Melting Pt: 208…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051886 CAS Number: 7484-37-9 EINECS: 231-289-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...