3-[1,1'-Biphenyl]-4-ylacrylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12643-14
A12643-09
A12643-18
CAAAAA12643-14EA
453.46
CAD
CAAAAA12643-14
CAAAA12643-09
CAAAA12643-18
3-[1,1'-Biphenyl]-4-ylacrylic acid 98%
3-[1,1'-Biphenyl]-4-ylacrylic acid
Purity: 98%
CAS No.: 13026-23-8
Molecular Formula: C15H12O2
Formula Weight: 224.27
Melting Point: 224-228°
Beilstein Reference: 1955124
MDL No.: MFCD00014010
Formula:
C₁₅H₁₂O₂ Melting Pt: 224…228 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014010 CAS Number: 13026-23-8 EINECS: 235-888-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...