DL-Alanine methyl ester hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12599-06
A12599-14
CAAAA12599-06EA
49.93
CAD
CAAAA12599-06
CAAAA12599-14
CAAAA12599-22
DL-Alanine methyl ester hydrochloride ≥98%
DL-Alanine methyl ester hydrochloride
Purity: 98+%
CAS No.: 13515-97-4
Molecular Formula: C4H9NO2.HCl
Formula Weight: 139.58
Storage Temperature: 2°C to 8°C
Melting Point: ca 155° dec.
Beilstein Reference: 3619264
MDL No.: MFCD00035523
Formula:
C₄H₉NO₂ Melting Pt: ca 155 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00035523 CAS Number: 13515-97-4 EINECS: 236-854-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...