4,6-Dichloro-o-toluidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4-Dichloro-6-methylaniline

A12591-06 A12591-14
CAAAAA12591-22EA 1355.06 CAD
CAAAAA12591-22 CAAAA12591-06 CAAAA12591-14
4,6-Dichloro-o-toluidine 97%
4,6-Dichloro-o-toluidine

Purity: 97% CAS No.: 30273-00-8 Molecular Formula: C7H7Cl2N Formula Weight: 176.05 Melting Point: 43-45° Boiling Point: 82-84°/0.1mm MDL No.: MFCD00044074

Formula: C₇H₇Cl₂N
Boiling Pt: 82…84 °C/0.1 mm
Melting Pt: 43…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00044074
CAS Number: 30273-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR