3-(Phenylthio)thiophene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12585-03
A12585-06
CAAAA12585-03EA
116.13
CAD
CAAAA12585-03
CAAAA12585-06
3-(Phenylthio)thiophene 97%
3-(Phenylthio)thiophene
Purity: 97%
CAS No.: 16718-11-9
Molecular Formula: C10H8S2
Formula Weight: 192.30
Boiling Point: 111-112°/0.6mm
MDL No.: MFCD00052043
Formula:
C₁₀H₈S₂ MW: 192.30 g/mol Boiling Pt: 111…112 °C/0.6 mm Storage Temperature: Ambient |
MDL Number:
MFCD00052043 CAS Number: 16718-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...