4-Chlorobenzenesulfinic acid sodium salt hydrate 97%

Supplier: Thermo Scientific Chemicals

A12583-06 A12583-14
CAAAA12583-06EA 93.38 CAD
CAAAA12583-06 CAAAA12583-14 CAAAA12583-22
4-Chlorobenzenesulfinic acid sodium salt hydrate 97%
4-Chlorobenzenesulfinic acid sodium salt hydrate

Purity: 97% CAS No.: 14752-66-0 Molecular Formula: C6H4ClNaO2S.xH2O Formula Weight: 198.60(anhy) Melting Point: 300° Beilstein Reference: 3575604 MDL No.: MFCD00035602

Formula: C₆H₄ClNaO₂S.xH₂O
MW: 198.60 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00035602
CAS Number: 14752-66-0
EINECS: 238-811-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR