Hydroxylamine-O-sulfonic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

A12560-14 A12560-22
CAAAA12560-14EA 165.9 CAD
CAAAA12560-14 CAAAA12560-22
Hydroxylamine-O-sulfonic acid 97%
Hydroxylamine-O-sulphonic acid

Fieser: 1,481 3,156 4,256 5,343 6,290 8,250 9,245 10,207 15,170


Purity: 97% CAS No.: 2950-43-8 Molecular Formula: NH2OSO3H Formula Weight: 113.09 Storage Temperature: 2°C to 8°C Melting Point: ca 210° dec. Beilstein Reference: 956566 UN No.: 3260 MDL No.: MFCD00011604

Formula: H₃NO₄S
Melting Pt: ∼ 210 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00011604
CAS Number: 2950-43-8
EINECS: 220-971-6
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR