3-Ethoxycarbonylcoumarin 98%
Supplier: Thermo Scientific Chemicals
A12553-03
A12553-06
CAAAA12553-03EA
39.34
CAD
CAAAA12553-03
CAAAA12553-06
3-Ethoxycarbonylcoumarin 98%
3-Ethoxycarbonylcoumarin
Purity: 98%
CAS No.: 1846-76-0
Molecular Formula: C12H10O4
Formula Weight: 218.21
Melting Point: 92-95°
Beilstein Reference: 200147
MDL No.: MFCD00016964
Formula:
C₁₂H₁₀O₄ Melting Pt: 92…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016964 CAS Number: 1846-76-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...