Penta-O-acetyl-β-D-glucopyranose 98%
Supplier: Thermo Scientific Chemicals
A12526-0B
A12526-18
A12526-30
CAAAA12526-0BEA
750.5
CAD
CAAAA12526-0B
CAAAA12526-18
CAAAA12526-30
Penta-O-acetyl-β-D-glucopyranose 98%
Penta-O-acetyl-beta-D-glucopyranose
Purity: 98%
CAS No.: 604-69-3
Molecular Formula: C16H22O11
Formula Weight: 390.34
Melting Point: 130-132°
Beilstein Reference: 98851
Optical Rotation: +5° (c=5 in chloroform)
MDL No.: MFCD00006597
Formula:
C₁₆H₂₂O₁₁ Melting Pt: 130…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006597 CAS Number: 604-69-3 EINECS: 210-074-8 |
Specification Test Results
Optical rotation | +5° (c=5 in chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...